missing translation for 'onlineSavingsMsg'
Learn More

3,6-Difluorophthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 11382059
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382059 1 g 1g
15453377 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382059 Supplier Thermo Scientific Alfa Aesar Supplier No. H26096.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 651-97-8
Molecular Formula C8H4F2O4
Molecular Weight (g/mol) 202.113
MDL Number MFCD03840499
InChI Key VFLMWMTWWZGXGA-UHFFFAOYSA-N
Synonym 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro
PubChem CID 643385
IUPAC Name 3,6-difluorophthalic acid
SMILES C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F

Specifications

Melting Point ∼184°C (decomposition)
Quantity 1 g
Formula Weight 202.11
Percent Purity 98%
Chemical Name or Material 3,6-Difluorophthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.