missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloropyridazine, 98%

Product Code. 11492527
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492527 100 g 100g
11482527 25 g 25g
11402537 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492527 Supplier Thermo Scientific Alfa Aesar Supplier No. A14795.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-30-0 | C4H2Cl2N2 | 148.974 g/mol

3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-30-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006466
InChI Key GUSWJGOYDXFJSI-UHFFFAOYSA-N
Synonym pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
PubChem CID 67331
IUPAC Name 3,6-dichloropyridazine
SMILES C1=CC(=NN=C1Cl)Cl

Specifications

Melting Point 66°C to 70°C
Quantity 100 g
UN Number UN2811
Beilstein 111151
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 148.98
Percent Purity 98%
Chemical Name or Material 3,6-Dichloropyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.