missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-4-methylpyridazine, 97%, Thermo Scientific™

Product Code. 10338930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338930 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10338930 Supplier Thermo Scientific Acros Supplier No. 305820010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 19064-64-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 84°C to 89°C
Color Brown
Boiling Point 149°C to 151°C (21.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H4Cl2N2
MDL Number MFCD00006465
Quantity 1 g
Synonym 4-methyl-3,6-dichloropyridazine, 3,6-dichloro-4-methyl-pyridazine, pyridazine, 3,6-dichloro-4-methyl, 3,6-dichloro-5-methylpyridazine, 3,6-dichloro-4-methyl-1,2-diazine, pubchem9497, ksc495e6l, 3,6-dichloro-4-methylpyridazin, pyridazine,6-dichloro-4-methyl, 3,6-di chloro-4-methyl pyridazine
InChI Key ROYHWGZNGMXQEU-UHFFFAOYSA-N
SMILES CC1=CC(=NN=C1Cl)Cl
IUPAC Name 3,6-dichloro-4-methylpyridazine
Molecular Weight (g/mol) 163.01
PubChem CID 87923
Formula Weight 163.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 6-Dichloro-4-methylpyridazine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.