missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-2-hydroxybenzoic Acid Potassium Salt, TRC

Product Code. 30380038
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380038 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380038 Supplier Toronto Research Chemicals Supplier No. TRCD434515100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1429059-62-0
Molecular Formula C7H3Cl2KO3
Quantity 100 mg
Synonym 3,6-dichloro-(6CI,7CI,8CI) Salicylic Acid, 3,6-Dichloro-2-hydroxybenzoic Acid Potassium Salt, 2-Hydroxy-3,6-dichlorobenzoic Acid Potassium Salt, 3,6-DCSA Potassium Salt, 3,6-Dichlorosalicylic Acid Potassium Salt
InChI Formula InChI=1S/C7H4Cl2O3.K/c8-3-1-2-4(9)6(10)5(3)7(11)12;/h1-2,10H,(H,11,12);/q;+1/p-1
SMILES O=C(O[K])C1=C(Cl)C=CC(Cl)=C1O
Molecular Weight (g/mol) 245.1
Chemical Name or Material 3,6-Dichloro-2-hydroxybenzoic Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.