missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromocarbazole, 99%

Product Code. 15422828
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422828 25 g 25g
15412828 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422828 Supplier Thermo Scientific Alfa Aesar Supplier No. H56360.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6825-20-3 | C12H7Br2N | 325.003 g/mol

3,6-Dibromocarbazole has been used in the preparation of N-(2-hydroxyethyl)-3,6-dibromocarbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dibromocarbazole has been used in the preparation of N-(2-hydroxyethyl)-3,6-dibromocarbazole.

Solubility
Soluble in benzene, chloroform

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6825-20-3
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325.003
MDL Number MFCD00004961
InChI Key FIHILUSWISKVSR-UHFFFAOYSA-N
Synonym 3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t
PubChem CID 274874
IUPAC Name 3,6-dibromo-9H-carbazole
SMILES C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br

Specifications

Melting Point 204°C to 206°C
Quantity 25 g
Solubility Information Soluble in benzene,chloroform
Formula Weight 325
Percent Purity 99%
Chemical Name or Material 3,6-Dibromocarbazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.