missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%, Thermo Scientific Chemicals

Product Code. 15420358
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1671-87-0
Molecular Formula C12H8N6
Molecular Weight (g/mol) 236.24
MDL Number MFCD00121717
InChI Key JFBIRMIEJBPDTQ-UHFFFAOYSA-N
Synonym 3,6-di pyridin-2-yl-1,2,4,5-tetrazine, 3,6-di 2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridinyl-1,2,4,5-tetrazine, 3,6-bis 2-pyridyl-1,2,4,5-tetrazine, bis pyridin-2-yl-1,2,4,5-tetrazine, 3,6-bis 2-pyridinyl-1,2,4,5-tetrazine, bptz, acmc-209dvt, 3,6-di-2-pyridyl-s-tetrazine
PubChem CID 315109
IUPAC Name 3,6-dipyridin-2-yl-1,2,4,5-tetrazine
SMILES C1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1

Specifications

Melting Point 225°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 236.24
Percent Purity 96%
Chemical Name or Material 3,6-Di-2-pyridyl-1,2,4,5-tetrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'