Learn More
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 349940050
Code : 29
Additional Details : CAS Number : 24748-23-0 Weight : 0.05500kg Transport : UN number : 3105 Chem class : 5.2
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsSpecifications
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane | |
41% Solution in Aromatic Free Min. Spirit | |
58.0 | |
60.0 | |
MFCD02685709 | |
unii-j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexoxonane, 1,2,4,5,7,8-hexoxonane, 3,6,9-triethyl-3,6,9-trimethyl, j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, unii-j3mv6azn38 component kvwlloiegklbpa-aqhstmncsa-n, unii-j3mv6azn38 component kvwlloiegklbpa-zsbigdgjsa-n, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxacyclononane, 1,2,4,5,7,8-hexaoxacyclononane, 3,6,9-trimethyl, 3,6,9-triethyl, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane sol.in arom.free min.spirit | |
KVWLLOIEGKLBPA-UHFFFAOYSA-N | |
3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane | |
2734078 | |
7.3 to 7.6% (total active oxygen) | |
0.875 | |
5 g | |
Solution |
Undesignated | |
24748-23-0,78-93-3 | |
<10.0°C | |
C12H24O6 | |
0.8750g/mL | |
Solubility in water: immiscible. Other solubilities: miscible with most organic solvents | |
CCC1(OOC(OOC(OO1)(C)CC)(C)CC)C | |
264.318 | |
264.32 | |
Plastic bottle | |
5 mPa.s (20°C) | |
74°C |
Safety and Handling
- 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, 41% solution in aromatic free mineral spirit
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Organic peroxide Type C & D
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P370+P380+P375-In case of fire: Evacuate area. Fight fire remotely due to the risk of explosion.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only