missing translation for 'onlineSavingsMsg'
Learn More

3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals

Product Code. 10718732 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 10718732

Brand: Thermo Scientific Acros 349941000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane
Color Undesignated
Name Note 41% Solution in Aromatic Free Min. Spirit
CAS 24748-23-0, 78-93-3
CAS Min % 40.0, 58.0
CAS Max % 42.0, 60.0
Melting Point <10.0°C
Molecular Formula C12H24O6
MDL Number MFCD02685709
Density 0.8750g/mL
Synonym unii-j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexoxonane, 1,2,4,5,7,8-hexoxonane, 3,6,9-triethyl-3,6,9-trimethyl, j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, unii-j3mv6azn38 component kvwlloiegklbpa-aqhstmncsa-n, unii-j3mv6azn38 component kvwlloiegklbpa-zsbigdgjsa-n, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxacyclononane, 1,2,4,5,7,8-hexaoxacyclononane, 3,6,9-trimethyl, 3,6,9-triethyl, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane sol.in arom.free min.spirit
Solubility Information Solubility in water: immiscible. Other solubilities: miscible with most organic solvents
InChI Key KVWLLOIEGKLBPA-UHFFFAOYSA-N
SMILES CCC1(OOC(OOC(OO1)(C)CC)(C)CC)C
IUPAC Name 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane
Molecular Weight (g/mol) 264.318
PubChem CID 2734078
Formula Weight 264.32
Percent Purity 7.3 to 7.6% (total active oxygen)
Packaging Plastic bottle
Specific Gravity 0.875
Viscosity 5 mPa.s (20°C)
Quantity 100 g
Flash Point 74°C
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, 41% solution in aromatic free mineral spirit
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Organic peroxide Type C & D
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P370+P380+P375-In case of fire: Evacuate area. Fight fire remotely due to the risk of explosion.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.