missing translation for 'onlineSavingsMsg'
Learn More

3,5-Xylenol Phosphate, TRC

Product Code. 30451791
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451791 1 g -
30449310 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451791 Supplier Toronto Research Chemicals Supplier No. TRCX7495501G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25653-16-1
Molecular Formula C24H27O4P
Molecular Weight (g/mol) 410.44
Synonym Tri-3,5-xylyl Phosphate, Phenol, 3,5-dimethyl-, 1,1',1' '-phosphate, 3,5-Xylenol, phosphate (3:1) (8CI), 3,5-Xylyl phosphate ((C8H9O)3PO) (6CI), 3,5-Xylyl phosphate (7CI), Phenol, 3,5-dimethyl-, phosphate (3:1) (9CI), NSC 66515, Tri-3,5-xylenyl phosphate, Tri-3,5-xylyl phosphate, Tris(3,5-dimethylphenyl) phosphate, Tri-3,5-xylyl phosphate, Tris(3,5-dimethylphenyl) Phosphate, 3,5-Dimethylphenol 1,1',1' '-Phosphate, 3,5-Dimethylphenol Phosphate (3:1)
IUPAC Name tris(3,5-dimethylphenyl) phosphate
SMILES Cc1cc(C)cc(OP(=O)(Oc2cc(C)cc(C)c2)Oc3cc(C)cc(C)c3)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H27O4P/c1-16-7-17(2)11-22(10-16)26-29(25,27-23-12-18(3)8-19(4)13-23)28-24-14-20(5)9-21(6)15-24/h7-15H,1-6H3
Formula Weight 410.1647
Percent Purity >95%
Chemical Name or Material 3,5-Xylenol Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.