missing translation for 'onlineSavingsMsg'
Learn More

3,5-Pyridinedicarboxylic Acid, TRC

Product Code. 30415968
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415968 10 g -
30423537 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415968 Supplier Toronto Research Chemicals Supplier No. TRCP99162010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-81-0
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
Synonym Dinicotinic Acid, 3,5-Pyridinedicarboxylate, 5-Carboxynicotinic Acid, NSC 6497
IUPAC Name pyridine-3,5-dicarboxylic acid
SMILES OC(=O)c1cncc(c1)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H5NO4/c9-6(10)4-1-5(7(11)12)3-8-2-4/h1-3H,(H,9,10)(H,11,12)
Formula Weight 167.0219
Chemical Name or Material 3,5-Pyridinedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.