missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Methyl-1H-pyrazol-4-yl)propylamine, Technical Grade, Thermo Scientific™

Product Code. 10061784
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061784 5 g Amber Glass Bottle 5g
10039591 250 mg Amber Glass Bottle 250mg
10642463 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10061784 Supplier Thermo Scientific Maybridge Supplier No. SEW05905DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 28739-42-6
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.202
InChI Key BYGCQGLHPCERCH-UHFFFAOYSA-N
Synonym 3-5-methyl-1h-pyrazol-4-yl propan-1-amine, 3-5-methyl-1h-pyrazol-4-yl propylamine, 3-3-methyl-2h-pyrazol-4-yl propan-1-amine, 4-3-aminopropyl-5-methylpyrazole, 3-5-methylpyrazol-4-yl propylamine, 1h-pyrazole-4-propanamine,3-methyl, 4-3-aminopropyl-5-methyl-1h-pyrazole, tech
PubChem CID 2736943
IUPAC Name 3-(5-methyl-1H-pyrazol-4-yl)propan-1-amine
SMILES CC1=C(C=NN1)CCCN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 139.2
Percent Purity 97%
Chemical Name or Material 3-(5-Methyl-1H-pyrazol-4-yl)propylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.