missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Lutidine, TRC
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
Specifications
Specifications
| Chemical Name or Material | 3,5-Lutidine |
| CAS | 591-22-0 |
| Molecular Formula | C7H9N |
| Quantity | 25 g |
| Synonym | 3,5-Dimethylpyridine, NSC 60736 |
| InChI Formula | InChI=1S/C7H9N/c1-6-3-7(2)5-8-4-6/h3-5H,1-2H3 |
| SMILES | Cc1cncc(C)c1 |
| IUPAC Name | 3,5-dimethylpyridine |
| Molecular Weight (g/mol) | 107.15 |
| Formula Weight | 107.0735 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction