missing translation for 'onlineSavingsMsg'
Learn More

3,5-Lutidine, 99%

Product Code. 11412677
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412677 500 mL 500mL
11359519 25 mL 25mL
11402677 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412677 Supplier Thermo Scientific Alfa Aesar Supplier No. A14877.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 591-22-0 | C7H9N | 107.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-22-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006404
InChI Key HWWYDZCSSYKIAD-UHFFFAOYSA-N
Synonym 3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825
PubChem CID 11565
IUPAC Name 3,5-dimethylpyridine
SMILES CC1=CC(=CN=C1)C

Specifications

Melting Point -9°C
Density 0.941
Boiling Point 169°C to 170°C
Flash Point 53°C (127°F)
Odor Unpleasant
Refractive Index 1.505
Quantity 500 mL
UN Number UN1992
Beilstein 105682
Solubility Information Sol
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material 3,5-Lutidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.