missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dioxocyclohexanecarboxylic Acid Ethyl Ester, TRC

Product Code. 30138451
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138451 2.5 g 2.5g
30004824 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138451 Supplier Toronto Research Chemicals Supplier No. TRCD4863702.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27513-35-5
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.19
Synonym Ethyl 3,5-Dioxocyclohexanecarboxylate
SMILES CCOC(=O)C1CC(=O)CC(=O)C1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H12O4/c1-2-13-9(12)6-3-7(10)5-8(11)4-6/h6H,2-5H2,1H3
Formula Weight 184.0736
Chemical Name or Material 3,5-Dioxocyclohexanecarboxylic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.