missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrosalicylaldehyde, 98%

Product Code. 11434367
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434367 1 g 1g
11444367 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434367 Supplier Thermo Scientific Alfa Aesar Supplier No. B20424.03
  • ONLINE EXCLUSIVE PRICE
    £31.30
    £28.65 / 1g
    Save £2.65
    8% Off
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2460-59-5 | C7H3N2O6 | 211.11 g/mol

It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

Solubility
Soluble in water, 0.6 g/100 mL (18°C).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2460-59-5
Molecular Formula C7H3N2O6
Molecular Weight (g/mol) 211.11
MDL Number MFCD00007103
InChI Key FLJXIBHYDIMYRS-UHFFFAOYSA-M
PubChem CID 75571
IUPAC Name 2-formyl-4,6-dinitrobenzen-1-olate
SMILES [O-]C1=C(C=O)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 71°C to 74°C
Quantity 1 g
Beilstein 1121900
Sensitivity Air Sensitive
Synonym 3,5-dinitrosalicylaldehyde, 2-hydroxy-3,5-dinitrobenzaldehyde, 3,5-dinitro-2-hydroxybenzaldehyde, benzaldehyde, 2-hydroxy-3,5-dinitro, acmc-20anby, 2-hydroxy-3,5-dinitro-benzaldehyde, 2-hydroxy-3,5-dinitrobenzaldehyde #, benzaldehyde,2-hydroxy-3,5-dinitro
Solubility Information Soluble in water,0.6g/100 mL (18°C).
Formula Weight 212.12
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrosalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.