missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzotrifluoride, 98%

Product Code. 11379839
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379839 100 g 100g
11359839 5 g 5g
11369839 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379839 Supplier Thermo Scientific Alfa Aesar Supplier No. B24248.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 401-99-0 | C7H3F3N2O4 | 236.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-99-0
Molecular Formula C7H3F3N2O4
Molecular Weight (g/mol) 236.106
MDL Number MFCD00007233
InChI Key QZADIXWDDVQVKM-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzotrifluoride, 1,3-dinitro-5-trifluoromethyl benzene, benzene, 1,3-dinitro-5-trifluoromethyl, 3,5-dinitrotrifluorotoluene, alpha,alpha,alpha-trifluoro-3,5-dinitrotoluene, pubchem10396, 3, 5-dinitrotrifluorotoluene, 3,5-dinitro-benzotrifluoride, 3,5-?dinitrobenzotrifluoride
PubChem CID 123060
IUPAC Name 1,3-dinitro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point 50°C to 52°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2058079
Formula Weight 236.11
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.