missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoic Acid, TRC

Product Code. 30386979
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize':
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30386979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30386979 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD4799055G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dinitrobenzoic Acid
CAS 99-34-3
Molecular Formula C7 H4 N2 O6
Quantity 5 g
Synonym 3,5-Dinitrobenzoic Acid, 3-Carboxy-1,5-dinitrobenzene, DNBA, NSC 8732
InChI Formula InChI=1S/C7H4N2O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H,(H,10,11)
SMILES OC(=O)c1cc(cc(c1)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 3,5-dinitrobenzoic acid
Molecular Weight (g/mol) 212.12
Formula Weight 212.0069
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.