missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitroaniline, 98%

Product Code. p-3211822 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10641365

Brand: Thermo Scientific Acros 116940010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-87-1 | C6H5N3O4 | 183.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-87-1
MDL Number MFCD00007263
InChI Key MPBZUKLDHPOCLS-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dinitro, aniline, 3,5-dinitro, unii-0ahr6k1n73, ccris 3109, 3,5-dinitrophenylamine, 3-5-dinitro-phenylamine, 3,5-dinitroanilin, aniline,5-dinitro, acmc-209mx8, benzenamine,3,5-dinitro
PubChem CID 12068
IUPAC Name 3,5-dinitroaniline
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 159°C to 163°C
Color Brown-Yellow to Yellow
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H5N3O4
Linear Formula (O2N)2C6H3NH2
Quantity 1 g
Beilstein 12, 759
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents,soluble in ethanol and diethylether
Molecular Weight (g/mol) 183.12
Formula Weight 183.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 5-Dinitroaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dinitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.