missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dinitro-o-toluic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 3,5-Dinitro-2-methyl benzoic acid |
| CAS | 28169-46-2 |
| Molecular Formula | C8 H6 N2 O6 |
| Quantity | 10 g |
| Synonym | 2-Methyl-3,5-dinitrobenzoic Acid, 3,5-Dinitro-2-methylbenzoic Acid, NSC 168527 |
| InChI Formula | InChI=1S/C8H6N2O6/c1-4-6(8(11)12)2-5(9(13)14)3-7(4)10(15)16/h2-3H,1H3,(H,11,12) |
| SMILES | Cc1c(cc(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)O |
| IUPAC Name | 2-methyl-3,5-dinitrobenzoic acid |
| Molecular Weight (g/mol) | 226.14 |
| Formula Weight | 226.0226 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction