missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitro-L-tyrosine Ammonium Salt, TRC

Product Code. 30386433
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386433 50 mg
30382304 100 mg
30376964 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386433 Supplier Toronto Research Chemicals Supplier No. TRCD48056050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9 H9 N3 O7 . x H3 N
Molecular Weight (g/mol) 288.21
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-dinitro-phenyl)propanoic acid;ammonia
SMILES N.N[C@@H](Cc1cc(c(O)c(c1)[N+](=O)[O-])[N+](=O)[O-])C(=O)O

Specifications

InChI Formula InChI=1S/C9H9N3O7.H3N/c10-5(9(14)15)1-4-2-6(11(16)17)8(13)7(3-4)12(18)19;/h2-3,5,13H,1,10H2,(H,14,15);1H3/t5-;/m0./s1
Quantity 50 mg
Shipping Condition Room Temperature
Chemical Name or Material 3,5-Dinitro-L-tyrosine Ammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.