missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylthiophenol, 97%

Product Code. 11318145
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318145 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11318145 Supplier Thermo Scientific Alfa Aesar Supplier No. L08403.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 38360-81-5 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38360-81-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00040226
InChI Key CESBAYSBPMVAEI-UHFFFAOYSA-N
Synonym 3,5-dimethylthiophenol, benzenethiol, 3,5-dimethyl, 3,5-dimethylbenzene-1-thiol, 3,5-dimethyl thiophenol, m-xylene-5-thiol, 5-mercapto-m-xylene, 3,5-xylenethiol, xylenethiol, mixed isomers, 3,5dimethylthiophenol, pubchem6885
PubChem CID 117548
IUPAC Name 3,5-dimethylbenzenethiol
SMILES CC1=CC(=CC(=C1)S)C

Specifications

Density 1.015
Boiling Point 127°C (50 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.5675
Quantity 25 g
UN Number UN2810
Beilstein 3233085
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylthiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.