Learn More
3,5-Dimethylpyrazin-2-amine, ≥97%, Maybridge
Brand: Maybridge BTB07911DA
Code : 31
Additional Details : CAS Number : 91678-81-8 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
91678-81-8 | |
123.159 | |
GZZRWBRYYZGTNW-UHFFFAOYSA-N | |
2774764 | |
CC1=CN=C(C(=N1)C)N |
C6H9N3 | |
MFCD00107172 | |
2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine | |
3,5-dimethylpyrazin-2-amine |
Specifications
1g | |
Amber Glass Bottle | |
96.5 | |
97+% | |
MFCD00107172 | |
GZZRWBRYYZGTNW-UHFFFAOYSA-N | |
3,5-dimethylpyrazin-2-amine | |
2774764 | |
97% |
3, 5-Dimethylpyrazin-2-amine | |
91678-81-8 | |
100.0 | |
C6H9N3 | |
2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine | |
CC1=CN=C(C(=N1)C)N | |
123.159 | |
123.16 |