missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylhydrazine hydrochloride, 97%

Product Code. 11433744
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433744 25 g 25g
11423744 5 g 5g
11443744 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433744 Supplier Thermo Scientific Alfa Aesar Supplier No. A11336.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60481-36-9 | C8H13ClN2 | 172.656 g/mol

3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60481-36-9
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00052269
InChI Key RSBQANBNDXZFIY-UHFFFAOYSA-N
Synonym 3,5-dimethylphenylhydrazine hydrochloride, 3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenylhydrazinehydrochloride, 3,5-dimethyl phenylhydrazine hydrochloride, 3,5-dimethylphenylhydrazine, hydrazine, 3,5-dimethylphenyl-, monohydrochloride, pubchem6407, ksc491e7r, 1-3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenyl hydrazine-hydrogen chloride 1/1
PubChem CID 12651341
IUPAC Name (3,5-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=CC(=CC(=C1)NN)C.Cl

Specifications

Melting Point 180°C (decomposition)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylphenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.