missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylacetonitrile, 98%

Product Code. 11358275
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358275 10 g 10g
11348275 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358275 Supplier Thermo Scientific Alfa Aesar Supplier No. L10170.09
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39101-54-7 | C10H11N | 145.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39101-54-7
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00060304
InChI Key LMUKNQSVBFEUKR-UHFFFAOYSA-N
Synonym 3,5-dimethylphenylacetonitrile, 2-3,5-dimethylphenyl acetonitrile, 3,5-dimethylphenyl acetonitrile, benzeneacetonitrile, 3,5-dimethyl, 3,5-dimethyl-phenyl-acetonitrile, 2-3,5-dimethylphenyl ethanenitrile, acmc-20a3vm, 3.5-dimethylphenylacetonitril, 3,5-dimethylbenzeneacetonitrile, 3,5-dimethylphenylethanenitrile
PubChem CID 123481
IUPAC Name 2-(3,5-dimethylphenyl)acetonitrile
SMILES CC1=CC(=CC(=C1)CC#N)C

Specifications

Melting Point 42°C to 45°C
Boiling Point 170°C (20 mmHg)
Quantity 10 g
UN Number UN3439
Beilstein 2573956
Formula Weight 145.2
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.