missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10019511
Click to view available options
Amber Glass Bottle; 10g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 40046-30-8
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H9NS
Linear Formula (CH3)2C6H3NCS
MDL Number MFCD00041082
Synonym 3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
InChI Key DSMXCADWIFIJEX-UHFFFAOYSA-N
SMILES CC1=CC(=CC(=C1)N=C=S)C
IUPAC Name 1-isothiocyanato-3,5-dimethylbenzene
Molecular Weight (g/mol) 163.238
PubChem CID 142406
Formula Weight 163.24
Percent Purity 97%
Quantity 10 g
Chemical Name or Material 3, 5-Dimethylphenyl isothiocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'