missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10361875
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10361875

Brand: Thermo Scientific Maybridge CC00601DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2510-36-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
InChI Key IJEUISLJVBUNRE-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555
PubChem CID 75636
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(=NO1)C)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 141.13
Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.