missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzyl bromide, 98%

Product Code. p-2690268 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10691263

Brand: Thermo Scientific Acros 355560050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27129-86-8 | C9H11Br | 199.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27129-86-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00013539
InChI Key QXDHXCVJGBTQMK-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzyl bromide, 1-bromomethyl-3,5-dimethylbenzene, 3,5-dimethylbenzylbromide, alpha-bromomesitylene, benzene, 1-bromomethyl-3,5-dimethyl, .alpha.-bromomesitylene, 5-bromomethyl-1,3-dimethylbenzene, a-bromomesitylene, pubchem9979, 3,5dimethylbenzyl bromide
PubChem CID 141334
IUPAC Name 1-(bromomethyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)CBr)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36°C to 40°C
Color Yellow
Boiling Point 101°C to 103°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 199.09
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3, 5-Dimethylbenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethylbenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.