missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzyl bromide, 98%

Product Code. 11412417
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412417 25 g 25g
11402417 5 g 5g
11422417 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412417 Supplier Thermo Scientific Alfa Aesar Supplier No. A14267.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27129-86-8 | C9H11Br | 199.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27129-86-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00013539
InChI Key QXDHXCVJGBTQMK-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzyl bromide, 1-bromomethyl-3,5-dimethylbenzene, 3,5-dimethylbenzylbromide, alpha-bromomesitylene, benzene, 1-bromomethyl-3,5-dimethyl, .alpha.-bromomesitylene, 5-bromomethyl-1,3-dimethylbenzene, a-bromomesitylene, pubchem9979, 3,5dimethylbenzyl bromide
PubChem CID 141334
IUPAC Name 1-(bromomethyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)CBr)C

Specifications

Melting Point 36°C to 40°C
Boiling Point 109°C to 110°C (14 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 1857343
Formula Weight 199.1
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylbenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.