missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzoyl chloride, TRC

Product Code. 30375742
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375742 10 g 10g
30371431 5 g -
30381595 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375742 Supplier Toronto Research Chemicals Supplier No. TRCD47675810G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6613-44-1
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
SMILES Cc1cc(C)cc(c1)C(=O)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H9ClO/c1-6-3-7(2)5-8(4-6)9(10)11/h3-5H,1-2H3
Formula Weight 168.0342
Chemical Name or Material 3,5-Dimethylbenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.