missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzoic acid, 98+%

Product Code. 11423504
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423504 10 g 10g
11433504 50 g 50g
11443504 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423504 Supplier Thermo Scientific Alfa Aesar Supplier No. A11198.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 499-06-9 | C9H10O2 | 150.177 g/mol

3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

Solubility
Soluble in methanol. (1 g/10 mL). Slightly soluble in water.

Notes
Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-06-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002525
InChI Key UMVOQQDNEYOJOK-UHFFFAOYSA-N
Synonym mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid
PubChem CID 10356
ChEBI CHEBI:64821
IUPAC Name 3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C

Specifications

Melting Point 168°C to 172°C
Quantity 10 g
Odor Odorless
Beilstein 1072182
Solubility Information Soluble in methanol. (1g/10 mL). Slightly soluble in water.
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.