missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzenesulfonyl chloride, 97%

Product Code. p-7047305
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11309435

Brand: Thermo Scientific Alfa Aesar L18603.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-27-3 | C8H9ClO2S | 204.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-27-3
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.67
MDL Number MFCD03094655
InChI Key LSAGRAXLOLZVKO-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzene-1-sulfonyl chloride, 3,5-dimethylphenyl chlorosulfone, 3,5-dimethylbenzenesulfonylchloride, 3,5-dimethylbenzenesulphonyl chloride, 3,5-dimethyl-benzenesulfonyl chloride, pubchem5129, acmc-1ccrc, 5-chlorosulphonyl-m-xylene, 3,5-dimethylphenylsulfonyl chloride, 3,5-xylenesulfonyl chloride
PubChem CID 2736235
SMILES CC1=CC(=CC(C)=C1)S(Cl)(=O)=O

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
UN Number UN3261
Beilstein 2640617
IUPAC Name 3,5-dimethylbenzenesulfonyl chloride
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylbenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.