missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10177243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177243 5 g Glass bottle 5g
10350294 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177243 Supplier Thermo Scientific Acros Supplier No. 391590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2905-27-3
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.67
MDL Number MFCD03094655
InChI Key LSAGRAXLOLZVKO-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzene-1-sulfonyl chloride, 3,5-dimethylphenyl chlorosulfone, 3,5-dimethylbenzenesulfonylchloride, 3,5-dimethylbenzenesulphonyl chloride, 3,5-dimethyl-benzenesulfonyl chloride, pubchem5129, acmc-1ccrc, 5-chlorosulphonyl-m-xylene, 3,5-dimethylphenylsulfonyl chloride, 3,5-xylenesulfonyl chloride
PubChem CID 2736235
IUPAC Name 3,5-dimethylbenzenesulfonyl chloride
SMILES CC1=CC(=CC(C)=C1)S(Cl)(=O)=O

Specifications

Melting Point 93°C to 95°C
CAS Min % 96.0
Color White to Yellow
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2C6H3SO2Cl
Formula Weight 200.64
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethylbenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.