missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylanisole, 99%

Product Code. 10053373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10053373 100 g Glass bottle 100g
10172620 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10053373 Supplier Thermo Scientific Acros Supplier No. 115961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 874-63-5 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-63-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00008398
InChI Key JCHJBEZBHANKGA-UHFFFAOYSA-N
Synonym 3,5-dimethylanisole, 5-methoxy-m-xylene, benzene, 1-methoxy-3,5-dimethyl, 1-methoxy-3,5-dimethyl-benzene, 3,5-dimethylanisol, 3,5-dimethylanizole, acmc-1bkxt, anisole, 3,5-dimethyl, ksc494e7h, 5-methoxy-1,3-dimethylbenzene
PubChem CID 70126
IUPAC Name 1-methoxy-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)OC)C

Specifications

CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Density 0.9600g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5117 to 1.5137
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.96
Linear Formula (CH3)2C6H3OCH3
Beilstein 06,493
Boiling Point 193°C
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethylanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.