missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylaniline, 97+%

Product Code. 11423224
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423224 100 g 100g
11433224 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423224 Supplier Thermo Scientific Alfa Aesar Supplier No. A10570.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-69-0 | C8H11N | 121.183 g/mol

3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant. 3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant. 3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography.

Solubility
Soluble in water. <0.1 g/100 mL at 19°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and light sensitive. Incompatible to acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates, halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-69-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007813
InChI Key MKARNSWMMBGSHX-UHFFFAOYSA-N
Synonym 3,5-xylidine, benzenamine, 3,5-dimethyl, 3,5-xylylamine, 3,5-dimethylphenylamine, 3,5-xylidene, 5-amino-1,3-xylene, 1-amino-3,5-dimethylbenzene, 3,5-dimethylbenzenamine, 3,5-dimethylbenzeneamine, 5-amino-1,3-dimethylbenzene
PubChem CID 7949
IUPAC Name 3,5-dimethylaniline
SMILES CC1=CC(=CC(=C1)N)C

Specifications

Melting Point 9°C to 11°C
Density 0.971
Quantity 100 g
Odor Characteristic
Refractive Index 1.558
UN Number UN1711
Beilstein 507051
Merck Index 14,10084
Boiling Point 220°C to 221°C
Solubility Information Soluble in water. <0.1g/100 mL at 19°C
Formula Weight 121.18
Percent Purity ≥97%
Chemical Name or Material 3,5-Dimethylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.