missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyladamantane-1-carboxylic acid, 97%

Product Code. 15403607
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15403607 Supplier Thermo Scientific Alfa Aesar Supplier No. H27073.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14670-94-1 | C13H19O2 | 207.29 g/mol

3,5-Dimethyladamantane-1-carboxylic acid, is used as a pharmaceutical and API intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyladamantane-1-carboxylic acid, is used as a pharmaceutical and API intermediate.

Solubility
Sparingly Soluble (0.14 g/L) (25°C)

Notes
Store in cool place. Keep container tightly close
TRUSTED_SUSTAINABILITY

Specifications

CAS 14670-94-1
Melting Point 107°C to 109°C
Molecular Formula C13H19O2
MDL Number MFCD00188067
Quantity 1 g
Synonym 3,5-dimethyl-1-adamantanecarboxylic acid, 1,3-dimethyladamantan-5-carboxylic acid, 3,5-dimethyladamantanecarboxylic acid, maybridge1_002240, 3,5-dimethyltricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3,5-dimethyl adamantane-1-carboxylic acid, acmc-20ancb, 3,5-dimethyladamantane carboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylicacid, 3,5-dimethyl
Solubility Information Sparingly Soluble (0.14g/L) (25°C)
InChI Key BSWOQWGHXZTDOO-QGVZIFMBSA-M
SMILES C[C@@]12CC3C[C@@](C)(C1)CC(C3)(C2)C([O-])=O
IUPAC Name 3,5-dimethyladamantane-1-carboxylic acid
Molecular Weight (g/mol) 207.29
PubChem CID 2736224
Formula Weight 208.3
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyladamantane-1-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.