missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-isoxazolecarbaldehyde, Thermo Scientific™

Product Code. 10384182
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10384182 1 g 1g
10437802 250 mg 250mg
10623795 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10384182 Supplier Thermo Scientific Maybridge Supplier No. CC00604DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54593-26-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
InChI Key TVAYXKLCEILMEA-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
PubChem CID 289576
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=C(C(=NO1)C)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-4-isoxazolecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.