missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-hydroxybenzonitrile, TRC

Product Code. 30376142
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376142 100 mg
30374000 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376142 Supplier Toronto Research Chemicals Supplier No. TRCD473480100MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4198-90-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.17
Synonym 4-Hydroxy-3,5-dimethyl-benzonitrile, 2,6-Dimethyl-4-cyanophenol, 4-Cyano-2,6-dimethylphenol, 4-Hydroxy-3,5-dimethylbenzonitrile
SMILES Cc1cc(cc(C)c1O)C#N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H9NO/c1-6-3-8(5-10)4-7(2)9(6)11/h3-4,11H,1-2H3
Formula Weight 147.0684
Chemical Name or Material 3,5-Dimethyl-4-hydroxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.