missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-hydroxybenzaldehyde, 99%, ACROS Organics™

Product Code. 10443073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10443073 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10443073 Supplier Thermo Scientific Acros Supplier No. 381850050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2233-18-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point 112°C to 116°C
Color Brown to Tan
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula (CH3)2C6H2(OH)CHO
MDL Number MFCD00006946
Quantity 5 g
Synonym 3,5-dimethyl-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-dimethyl, 4-formyl-2,6-xylenol, 4-hydroxy-3,5-dimethyl-benzaldehyde, unii-23pa2pmp9f, 23pa2pmp9f, syringnldehyde, zlchem 510, pubchem8252, buttpark 9657-68
InChI Key UYGBSRJODQHNLQ-UHFFFAOYSA-N
SMILES CC1=CC(C=O)=CC(C)=C1O
IUPAC Name 4-hydroxy-3,5-dimethylbenzaldehyde
Molecular Weight (g/mol) 150.18
PubChem CID 75222
Formula Weight 150.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.