missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-3-hexanol, 98%, Thermo Scientific™

Product Code. 11470671
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11470671 1 g 1g
11480671 5 g 5g
11490671 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11470671

Brand: Thermo Scientific Alfa Aesar B20141.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4209-91-0
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00021823
InChI Key INMGJWCKWKKMPN-UHFFFAOYNA-N
Synonym 3,5-dimethyl-3-hexanol, 3-hexanol, 3,5-dimethyl, acmc-209jnj, 3-hexanol,3,5-dimethyl, 3-hexanol, 3,5-dimethyl-, .+/-.
PubChem CID 98266
IUPAC Name 3,5-dimethylhexan-3-ol
SMILES CCC(C)(O)CC(C)C

Specifications

Density 0.83
Quantity 1 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3,5-Dimethyl-3-hexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.