missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrrole-2-carbaldehyde, ≥95%, Thermo Scientific™

Product Code. 10580552
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10580552 1 g 1g
10069101 250 mg 250mg
10417174 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10580552 Supplier Thermo Scientific Maybridge Supplier No. CC18004DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2199-58-8
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00111522
InChI Key RDFZYUOHJBXMJA-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole
PubChem CID 270465
IUPAC Name 3,5-dimethyl-1H-pyrrole-2-carbaldehyde
SMILES CC1=CC(=C(N1)C=O)C

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula Yes
Quantity 1 g
Formula Weight 123.15
Percent Purity 95%
Chemical Name or Material 3, 5-Dimethyl-1H-pyrrole-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.