missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride, 97%

Product Code. 15420478
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420478 5 g 5g
15480696 250 mg 250mg
15410478 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420478 Supplier Thermo Scientific Alfa Aesar Supplier No. H54050.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80466-78-0 | C5H7ClN2O2S | 194.63 g/mol

3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80466-78-0
Molecular Formula C5H7ClN2O2S
Molecular Weight (g/mol) 194.63
MDL Number MFCD04969910
InChI Key NBGSZCQLPLYKJH-UHFFFAOYSA-N
Synonym 1h-pyrazole-4-sulfonylchloride, 3,5-dimethyl, 3,5-dimethylpyrazol-4-yl chlorosulfone, 1h-pyrazole-4-sulfonyl chloride, 3,5-dimethyl, pyrazole, 4-chlorosulfonyl-3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-sulfonylchloride, #
PubChem CID 594587
IUPAC Name 3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride
SMILES CC1=C(C(C)=NN1)S(Cl)(=O)=O

Specifications

Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 194.64
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.