missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10683435
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10683435 10 g Amber Glass Bottle 10g
10496082 250 mg Amber Glass Bottle 250mg
10793433 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683435 Supplier Thermo Scientific Maybridge Supplier No. MO00744EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61226-19-5
Molecular Formula C12H11N2O2
Molecular Weight (g/mol) 215.23
MDL Number MFCD02656670
InChI Key LPYTYYLNGJGJGW-UHFFFAOYSA-M
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid,3,5-dimethyl-1-phenyl, 1h-pyrazole-4-carboxylicacid, 3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbox ylic acid
PubChem CID 612275
SMILES CC1=NN(C(C)=C1C([O-])=O)C1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 3,5-dimethyl-1-phenylpyrazole-4-carboxylic acid
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.