missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15445145
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445145 250 mg 250mg
15435145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445145 Supplier Thermo Scientific Alfa Aesar Supplier No. H32478.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126779-11-0
Molecular Formula C16H27BN2O3
Molecular Weight (g/mol) 306.213
MDL Number MFCD12031452
InChI Key ZNDYJQBCSOGOJQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-tetrahydro-2h-pyran-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-thp-3,5-dimethylpyrazole-4-boronic acid pinacol ester, 3,5-dimethyl-1-2-tetrahydropyranyl-1h-pyrazole-4-boronic acid pinacol ester, 3,5-dimethyl-1-oxan-2-yl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3,5-dimethyl-1-oxan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-thp-3,5-dimethylpyrazole-4-boronic acid, pinacol ester, 3,5-dimethyl-1-oxan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3,5-dimethyl-1-tetrahydro-pyran-2-yl-1h-pyrazoleboronic acid pinacol ester, 3,5-dimethyl-1-tetrahydro-2h-pyran-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol
PubChem CID 53217144
IUPAC Name 3,5-dimethyl-1-(oxan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N(N=C2C)C3CCCCO3)C

Specifications

Quantity 250 mg
Formula Weight 306.22
Percent Purity 95%
Chemical Name or Material 3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.