missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenol, TRC

Product Code. 30009589
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009589 10 g 10g
30011307 100 g 100g
30004741 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009589 Supplier Toronto Research Chemicals Supplier No. TRCD46101510G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-99-2
Molecular Formula C8 H10 O3
Molecular Weight (g/mol) 154.16
Synonym 1-Hydroxy-3,5-dimethoxybenzene, NSC 70955, Phloroglucinol Dimethyl Ether, Taxicatigenin;
IUPAC Name 3,5-dimethoxyphenol
SMILES COc1cc(O)cc(OC)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H10O3/c1-10-7-3-6(9)4-8(5-7)11-2/h3-5,9H,1-2H3
Formula Weight 154.063
Chemical Name or Material 3,5-Dimethoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.