missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenol, 97%

Product Code. 10235680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235680 10 g Glass bottle 10g
10341474 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Produktkod. 10235680 Leverantör Thermo Scientific Acros Leverantörsnummer 158120100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 500-99-2 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 500-99-2
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008388
InChI Key XQDNFAMOIPNVES-UHFFFAOYSA-N
Synonym taxicatigenin, phenol, 3,5-dimethoxy, phloroglucinol dimethyl ether, 1-hydroxy-3,5-dimethoxybenzene, 3,5-dimethoxy phenol, unii-23uxw8136a, phenol,5-dimethoxy, 3,5-dimethoxy-phenol, pubchem16421, acmc-209kk2
PubChem CID 10383
IUPAC Name 3,5-dimethoxyphenol
SMILES COC1=CC(OC)=CC(O)=C1

Specifikationer

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color Brown-Pink to Colorless
Boiling Point 172°C to 175°C (17.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3OH
Quantity 10 g
Beilstein 06, 1101
Formula Weight 154.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Dimethoxyphenol
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 3,5-Dimethoxyphenol
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.