missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzylamine, 98%, ACROS Organics™

Product Code. 11601409 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11601409 5 g 5g
11925311 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11601409 Supplier Thermo Scientific Acros Supplier No. 437800050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 34967-24-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
InChI Key YGZJTYCCONJJGZ-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine
PubChem CID 420973
IUPAC Name (3,5-dimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1)CN)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or White
Density 1.11g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 167.21
Percent Purity ≥97.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3, 5-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.