missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzeneboronic acid, 98%

Product Code. 15409737
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409737 1 g 1g
15419737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409737 Supplier Thermo Scientific Alfa Aesar Supplier No. H52965.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 192182-54-0 | C8H11BO4 | 181.982 g/mol

3,5-Dimethoxybenzeneboronic acid is used as reactant for palladium-catalyzed coupling reactions, preparation of benzopyranone derivatives as positive GABAA receptor modulators, preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling. It is also used as reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide and preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzeneboronic acid is used as reactant for palladium-catalyzed coupling reactions, preparation of benzopyranone derivatives as positive GABAA receptor modulators, preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling. It is also used as reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide and preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192182-54-0
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD03095127
InChI Key XUIURRYWQBBCCK-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzeneboronic acid, 3,5-dimethoxyphenyl boronic acid, 3,5-dimethoxyphenylboronicacid, boronic acid, 3,5-dimethoxyphenyl, pubchem1827, acmc-1c82c, ksc174i6h, 3,5-dimethoxy phenyl boronic acid, 3,5-dimethoxyphenylboronic acid, boronic acid, b-3,5-dimethoxyphenyl
PubChem CID 4374257
IUPAC Name (3,5-dimethoxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)OC)OC)(O)O

Specifications

Melting Point 205°C to 207°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.