missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzaldehyde, 98%

Product Code. 11482058
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482058 100 g 100g
11462058 5 g 5g
11472058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482058 Supplier Thermo Scientific Alfa Aesar Supplier No. A15109.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7311-34-4 | C9H10O3 | 166.18 g/mol

3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7311-34-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00003366
InChI Key VFZRZRDOXPRTSC-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-benzaldehyde, benzaldehyde, 3,5-dimethoxy, 3,5-dimethoxybenzaldehyd, pubchem2645, 3,5 dimethoxybenzaldehyde, acmc-1bhx6, 3,5 dimethoxylbenzaldehyde, 3,5 dimethoxy benzaldehyde, 3,5-dimethoxy benzaldehyde, ghl.pd_mitscher_leg0.694
PubChem CID 81747
IUPAC Name 3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 115°C to 118°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2042374
Sensitivity Air Sensitive
Solubility Information Soluble in hot methanol. Insoluble in water.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.