missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyaniline, TRC

Product Code. 30387432
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387432 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387432 Supplier Toronto Research Chemicals Supplier No. TRCD4605905G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dimethoxyaniline
CAS 10272-07-8
Molecular Formula C8H11NO2
Quantity 5 g
Synonym 3,5-Dimethoxy-benzenamine, 3,5-Dimethoxybenzenamine, 3,5-Dimethoxyphenylamine
InChI Formula InChI=1S/C8H11NO2/c1-10-7-3-6(9)4-8(5-7)11-2/h3-5H,9H2,1-2H3
SMILES COc1cc(N)cc(OC)c1
IUPAC Name 3,5-dimethoxyaniline
Molecular Weight (g/mol) 153.18
Formula Weight 153.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.