missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%

Product Code. 10031390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031390 25 g Glass bottle 25g
10386730 5 g Glass bottle 5g
10066183 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10031390 Supplier Thermo Scientific Acros Supplier No. 132880250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 134-96-3 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-96-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00006943
InChI Key KCDXJAYRVLXPFO-UHFFFAOYSA-N
Synonym syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
PubChem CID 8655
ChEBI CHEBI:67380
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.0°C to 113.0°C
Color Brown to Green-Yellow
Density 1.0130g/mL
Boiling Point 192.0°C to 193.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9145
Specific Gravity 1.013
Solubility Information Solubility in water: insoluble. Other solubilities: very sparingly soluble in petr ether, soluble in alcohol, ether, chloroform, hot benzene, glacial acetic acid
Formula Weight 182.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxy-4-hydroxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethoxy-4-hydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.